accessibility menu, dialog, popup

UserName

Viewing profile as user:

CAS RN 13820-40-1

IUPAC Name: palladium(2+) diammonium tetrachloride
Synonyms: palladium(2+) diammonium tetrachloride
Molecular Weight (g/mol): 284.3
Molecular Formula: Cl4H8N2Pd
InChi Key: IRKVTUVFHGUMMN-UHFFFAOYSA-L
SMILES: [NH4+].[NH4+].[Cl-].[Cl-].[Cl-].[Cl-].[Pd++]
Molecular Weight (g/mol) 
  • (3)
  • (2)
Quantity 
  • (1)
  • (1)
  • (3)
Percent Purity 
  • (1)
  • (1)
  • (2)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results