accessibility menu, dialog, popup

UserName

Viewing profile as user:
CH3OSiH2NOH3COH3C

CAS RN 13822-56-5

IUPAC Name: (3-aminopropyl)trimethoxysilane
Synonyms: (3-aminopropyl)trimethoxysilane
Molecular Weight (g/mol): 179.29
Molecular Formula: C6H17NO3Si
InChi Key: SJECZPVISLOESU-UHFFFAOYSA-N
SMILES: CO[Si](CCCN)(OC)OC
Boiling Point 
  • (3)
  • (3)
  • (3)
  • (3)
Molecular Weight (g/mol) 
  • (6)
  • (6)
Percent Purity 
  • (3)
  • (3)
  • (6)
Quantity 
  • (1)
  • (3)
  • (1)
  • (1)
  • (1)
  • (1)
  • (1)
  • (3)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (6)
  • (3)
  • (3)
  • (6)
  • (3)
  • (3)
  • (3)
  • (3)
  • (6)
  • (6)
  • (3)
  • (3)
  • (6)
  • (1)
  • (3)
  • (1)
  • (1)
  • (1)
  • (1)
  • (1)
  • (3)
  • (6)
  • (2)