accessibility menu, dialog, popup

UserName

Viewing profile as user:

CAS RN 13826-94-3

IUPAC Name: platinum(2+) dipotassium tetrabromide
Synonyms: platinum(2+) dipotassium tetrabromide
Molecular Weight (g/mol): 592.9
Molecular Formula: Br4K2Pt
InChi Key: AXZAYXJCENRGIM-UHFFFAOYSA-J
SMILES: [K+].[K+].[Br-].[Br-].[Br-].[Br-].[Pt++]
Physical Form 
  • (2)
Quantity 
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results