accessibility menu, dialog, popup

UserName

Viewing profile as user:
H3CSiClClCl

CAS RN 13829-21-5

IUPAC Name: trichloro(decyl)silane
Synonyms: trichloro(decyl)silane
Molecular Weight (g/mol): 275.71
Molecular Formula: C10H21Cl3Si
InChi Key: HLWCOIUDOLYBGD-UHFFFAOYSA-N
SMILES: CCCCCCCCCC[Si](Cl)(Cl)Cl
Percent Purity 
  • (1)
  • (2)
Quantity 
  • (1)
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results