accessibility menu, dialog, popup

UserName

Viewing profile as user:

CAS RN 13831-31-7

IUPAC Name: 2-chloro-2-oxoethyl acetate
Synonyms: 2-chloro-2-oxoethyl acetate
Molecular Weight (g/mol): 136.53
Molecular Formula: C4H5ClO3
InChi Key: HZDNNJABYXNPPV-UHFFFAOYSA-N
SMILES: CC(=O)OCC(Cl)=O
Boiling Point 
  • (2)
  • (4)
  • (2)
Molecular Weight (g/mol) 
  • (4)
  • (4)
Percent Purity 
  • (2)
  • (6)
Quantity 
  • (1)
  • (4)
  • (3)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (4)
  • (2)
  • (2)
  • (4)
  • (2)
  • (4)
  • (2)
  • (4)
  • (4)
  • (2)
  • (6)
  • (1)
  • (4)
  • (3)
  • (4)