accessibility menu, dialog, popup

UserName

Viewing profile as user:
H3COO(R)H3C(S)CH3(S)CH3O(R)(S)H3C(S)OHCH3CH3(R)CH3(S)

CAS RN 13832-70-7

IUPAC Name: octadecyl (2R,4aS,6aS,6bR,10S,12aS,12bR,14bS)-10-hydroxy-2,4a,6a,6b,9,9,12a-heptamethyl-13-oxo-1,2,3,4,4a,5,6,6a,6b,7,8,8a,9,10,11,12,12a,12b,13,14b-icosahydropicene-2-carboxylate
Synonyms: stearyl glycyrrhetinate
Molecular Weight (g/mol): 723.18
Molecular Formula: C48H82O4
InChi Key: WNIFXKPDILJURQ-WOJAWXFMSA-N
SMILES: CCCCCCCCCCCCCCCCCCOC(=O)[C@]1(C)CC[C@]2(C)CC[C@]3(C)C(=CC(=O)[C@@H]4[C@@]5(C)CC[C@H](O)C(C)(C)C5CC[C@@]34C)[C@H]2C1
Molecular Weight (g/mol) 
  • (1)
  • (2)
Quantity 
  • (1)
  • (1)
  • (1)
Percent Purity 
  • (2)
Physical Form 
  • (2)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (1)
  • (2)
  • (1)
  • (2)
  • (1)
  • (1)
  • (1)
  • (2)
  • (2)