accessibility menu, dialog, popup

UserName

Viewing profile as user:

CAS RN 138372-67-5

IUPAC Name: 2-(4-tert-butylphenyl)-5-{3-[5-(4-tert-butylphenyl)-1,3,4-oxadiazol-2-yl]phenyl}-1,3,4-oxadiazole
Synonyms: 2-(4-tert-butylphenyl)-5-{3-[5-(4-tert-butylphenyl)-1,3,4-oxadiazol-2-yl]phenyl}-1,3,4-oxadiazole
Molecular Weight (g/mol): 478.6
Molecular Formula: C30H30N4O2
InChi Key: FQJQNLKWTRGIEB-UHFFFAOYSA-N
SMILES: CC(C)(C)C1=CC=C(C=C1)C1=NN=C(O1)C1=CC(=CC=C1)C1=NN=C(O1)C1=CC=C(C=C1)C(C)(C)C
Percent Purity 
  • (2)
Physical Form 
  • (2)
Quantity 
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results