CAS RN 1384281-49-5
IUPAC Name:
9-benzyl-2,7-dibromo-9H-carbazole
Synonyms:
9-benzyl-2,7-dibromocarbazole
Molecular Weight (g/mol):
415.13
Molecular Formula:
C19H13Br2N
InChi Key:
HJQQHWYCZFAJHY-UHFFFAOYSA-N
SMILES:
BrC1=CC2=C(C=C1)C1=C(C=C(Br)C=C1)N2CC1=CC=CC=C1
1
–
1
of
1
results
1
9-Benzyl-2,7-dibromo-9H-carbazole 98.0+%, TCI America™
CAS: 1384281-49-5 Molecular Formula: C19H13Br2N Molecular Weight (g/mol): 415.128 InChI Key: HJQQHWYCZFAJHY-UHFFFAOYSA-N PubChem CID: 50936718 IUPAC Name: 9-benzyl-2,7-dibromocarbazole SMILES: C1=CC=C(C=C1)CN2C3=C(C=CC(=C3)Br)C4=C2C=C(C=C4)Br