accessibility menu, dialog, popup

UserName

Viewing profile as user:
ONN

CAS RN 138479-47-7

IUPAC Name: 2-(1H-pyrazol-1-yl)benzaldehyde
Synonyms: 2-(pyrazol-1-yl)benzaldehyde
Molecular Weight (g/mol): 172.19
Molecular Formula: C10H8N2O
InChi Key: KIGZYDJKBTYFBZ-UHFFFAOYSA-N
SMILES: O=CC1=CC=CC=C1N1C=CC=N1

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results