accessibility menu, dialog, popup

UserName

Viewing profile as user:

CAS RN 1384974-37-1

IUPAC Name: (4-methylbenzenesulfonyl)[2-({2-[(4-methylphenyl)methoxy]ethyl}amino)-1,2-diphenylethyl]azanide; chlororutheniumylium
Synonyms: (4-methylbenzenesulfonyl)[2-({2-[(4-methylphenyl)methoxy]ethyl}amino)-1,2-diphenylethyl]azanide; chlororutheniumylium
Molecular Weight (g/mol): 650.2
Molecular Formula: C31H33ClN2O3RuS
InChi Key: INKUCOHLIHBSDN-UHFFFAOYNA-M
SMILES: Cl[Ru+].CC1=CC=C(COCCNC(C([N-]S(=O)(=O)C2=CC=C(C)C=C2)C2=CC=CC=C2)C2=CC=CC=C2)C=C1
Physical Form 
  • (2)
Quantity 
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results