accessibility menu, dialog, popup

UserName

Viewing profile as user:

CAS RN 138500-85-3

IUPAC Name: 2-[4-(bromomethyl)phenyl]-4,4,5,5-tetramethyl-1,3,2-dioxaborolane
Synonyms: 2-[4-(bromomethyl)phenyl]-4,4,5,5-tetramethyl-1,3,2-dioxaborolane
Molecular Weight (g/mol): 297
Molecular Formula: C13H18BBrO2
InChi Key: CBUOGMOTDGNEAW-UHFFFAOYSA-N
SMILES: CC1(C)OB(OC1(C)C)C1=CC=C(CBr)C=C1
Molecular Weight (g/mol) 
  • (2)
  • (4)
Quantity 
  • (3)
  • (3)
Percent Purity 
  • (2)
  • (2)
Physical Form 
  • (2)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (2)
  • (2)
  • (2)
  • (2)
  • (2)
  • (4)
  • (3)
  • (3)
  • (2)
  • (2)
  • (2)