accessibility menu, dialog, popup

UserName

Viewing profile as user:
HNOPHNOP

CAS RN 138517-61-0

IUPAC Name: 2-(diphenylphosphanyl)-N-{2-[2-(diphenylphosphanyl)benzamido]cyclohexyl}benzamide
Synonyms: trost ligand
Molecular Weight (g/mol): 690.76
Molecular Formula: C44H40N2O2P2
InChi Key: AXMSEDAJMGFTLR-UHFFFAOYNA-N
SMILES: O=C(NC1CCCCC1NC(=O)C1=CC=CC=C1P(C1=CC=CC=C1)C1=CC=CC=C1)C1=CC=CC=C1P(C1=CC=CC=C1)C1=CC=CC=C1
Quantity 
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results