accessibility menu, dialog, popup

UserName

Viewing profile as user:

CAS RN 138517-66-5

IUPAC Name: tetracyclo[6.6.2.0²,⁷.0⁹,¹⁴]hexadeca-2,4,6,9,11,13-hexaene-15,16-diamine
Synonyms: tetracyclo[6.6.2.0²,⁷.0⁹,¹⁴]hexadeca-2,4,6,9,11,13-hexaene-15,16-diamine
Molecular Weight (g/mol): 236.32
Molecular Formula: C16H16N2
InChi Key: NWDYSRZJOLDMRE-UHFFFAOYNA-N
SMILES: NC1C(N)C2C3=CC=CC=C3C1C1=CC=CC=C21
Percent Purity 
  • (2)
Physical Form 
  • (2)
Quantity 
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (2)
  • (2)
  • (2)
  • (1)
  • (1)