accessibility menu, dialog, popup

UserName

Viewing profile as user:
CH3SHNNOON

CAS RN 138564-59-7

IUPAC Name: 5-methyl-2-[(2-nitrophenyl)amino]thiophene-3-carbonitrile
Synonyms: 5-methyl-2-[(2-nitrophenyl)amino]thiophene-3-carbonitrile
Molecular Weight (g/mol): 259.28
Molecular Formula: C12H9N3O2S
InChi Key: NPXUFPFFHANGDL-UHFFFAOYSA-N
SMILES: CC1=CC(C#N)=C(NC2=CC=CC=C2[N+]([O-])=O)S1
Percent Purity 
  • (2)
Physical Form 
  • (2)
Quantity 
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results