accessibility menu, dialog, popup

UserName

Viewing profile as user:

CAS RN 138642-97-4

NNClOOH

CAS RN 138642-97-4

IUPAC Name: 6-chloroimidazo[1,2-a]pyridine-3-carboxylic acid
Synonyms: 6-chloroimidazo[1,2-a]pyridine-3-carboxylic acid
Molecular Weight (g/mol): 196.59
Molecular Formula: C8H5ClN2O2
InChi Key: LXRDUEDQFSBROM-UHFFFAOYSA-N
SMILES: OC(=O)C1=CN=C2C=CC(Cl)=CN12

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results