accessibility menu, dialog, popup

UserName

Viewing profile as user:
H3CONHNHCl

CAS RN 1386928-34-2

IUPAC Name: 1-{4'-methoxy-[1,1'-biphenyl]-2-yl}piperazine hydrochloride
Synonyms: 1-{4'-methoxy-[1,1'-biphenyl]-2-yl}piperazine hydrochloride
Molecular Weight (g/mol): 304.82
Molecular Formula: C17H21ClN2O
InChi Key: KXZUJPBDJNUNCN-UHFFFAOYSA-N
SMILES: Cl.COC1=CC=C(C=C1)C1=CC=CC=C1N1CCNCC1

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results