accessibility menu, dialog, popup

UserName

Viewing profile as user:

CAS RN 13874-02-7

IUPAC Name: sodium tetrachlorogolduide dihydrate
Synonyms: sodium dihydrate tetrachloroaurate
Molecular Weight (g/mol): 397.79
Molecular Formula: AuCl4H4NaO2
InChi Key: OEERDTKOVZFOHF-UHFFFAOYSA-J
SMILES: O.O.[Na+].Cl[Au-](Cl)(Cl)Cl
Molecular Weight (g/mol) 
  • (2)
  • (2)
Quantity 
  • (2)
  • (2)
Physical Form 
  • (2)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results