accessibility menu, dialog, popup

UserName

Viewing profile as user:

CAS RN 138774-94-4

IUPAC Name: (2R)-2-({[(9H-fluoren-9-yl)methoxy]carbonyl}amino)-3-(naphthalen-2-yl)propanoic acid
Synonyms: (2R)-2-{[(9H-fluoren-9-ylmethoxy)carbonyl]amino}-3-(naphthalen-2-yl)propanoic acid|FMOC-3-(2-NAPHTHYL)-D-ALANINE
Molecular Weight (g/mol): 437.5
Molecular Formula: C28H23NO4
InChi Key: JYUTZJVERLGMQZ-AREMUKBSSA-N
SMILES: OC(=O)[C@@H](CC1=CC2=CC=CC=C2C=C1)NC(=O)OCC1C2=CC=CC=C2C2=CC=CC=C12
Quantity 
  • (2)
  • (1)
Grade 
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (2)
  • (1)
  • (2)
  • (2)
  • (1)
  • (1)