accessibility menu, dialog, popup

UserName

Viewing profile as user:

CAS RN 13880-05-2

IUPAC Name: N-[1,3-dihydroxy-2-(hydroxymethyl)propan-2-yl]prop-2-enamide
Synonyms: N-[1,3-dihydroxy-2-(hydroxymethyl)propan-2-yl]prop-2-enamide
Molecular Weight (g/mol): 175.18
Molecular Formula: C7H13NO4
InChi Key: MVBJSQCJPSRKSW-UHFFFAOYSA-N
SMILES: OCC(CO)(CO)NC(=O)C=C
Percent Purity 
  • (1)
  • (3)
Quantity 
  • (1)
  • (2)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results