accessibility menu, dialog, popup

UserName

Viewing profile as user:

CAS RN 13880-89-2

NNOH

CAS RN 13880-89-2

IUPAC Name: 3-hydroxypentanedinitrile
Synonyms: 3-hydroxypentanedinitrile
Molecular Weight (g/mol): 110.12
Molecular Formula: C5H6N2O
InChi Key: NMFITULDMUZCQD-UHFFFAOYSA-N
SMILES: OC(CC#N)CC#N
Percent Purity 
  • (2)
Quantity 
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (2)
  • (2)
  • (2)
  • (2)
  • (1)
  • (1)