accessibility menu, dialog, popup

UserName

Viewing profile as user:

CAS RN 138872-44-3

ONHSSOOCl

CAS RN 138872-44-3

IUPAC Name: 5-[(phenylformamido)methyl]thiophene-2-sulfonyl chloride
Synonyms: 5-[(phenylformamido)methyl]thiophene-2-sulfonyl chloride
Molecular Weight (g/mol): 315.79
Molecular Formula: C12H10ClNO3S2
InChi Key: VGSWVDWOXYTAPG-UHFFFAOYSA-N
SMILES: ClS(=O)(=O)C1=CC=C(CNC(=O)C2=CC=CC=C2)S1

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results