accessibility menu, dialog, popup

UserName

Viewing profile as user:
CH3ClOOCH3

CAS RN 13888-03-4

IUPAC Name: methyl 4-chloro-2-methylbutanoate
Synonyms: methyl 4-chloro-2-methylbutanoate
Molecular Weight (g/mol): 150.6
Molecular Formula: C6H11ClO2
InChi Key: ZDDYJHVTUSUPFC-UHFFFAOYNA-N
SMILES: COC(=O)C(C)CCCl
Percent Purity 
  • (2)
Quantity 
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results