accessibility menu, dialog, popup

UserName

Viewing profile as user:

CAS RN 138890-62-7

IUPAC Name: (4R)-4-(ethylamino)-2-(3-methoxypropyl)-1,1-dioxo-2H,3H,4H-1λ⁶-thieno[3,2-e][1,2]thiazine-6-sulfonamide
Synonyms: brinzolamide azopt brinzolamide (product) brinzolamide preparation brinzolamide (substance) allphar brand of brinzolamide alcon brand of brinzolamide
Molecular Weight (g/mol): 383.5
Molecular Formula: C12H21N3O5S3
InChi Key: HCRKCZRJWPKOAR-JTQLQIEISA-N
SMILES: CCN[C@H]1CN(CCCOC)S(=O)(=O)C2=C1C=C(S2)S(N)(=O)=O
Percent Purity 
  • (2)
Physical Form 
  • (2)
Quantity 
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results