accessibility menu, dialog, popup

UserName

Viewing profile as user:

CAS RN 13895-38-0

IUPAC Name: 5-methoxy-2-nitrosophenol
Synonyms: 5-methoxy-2-nitrosophenol
Molecular Weight (g/mol): 153.14
Molecular Formula: C7H7NO3
InChi Key: RNBFLAJNUZQUIK-UHFFFAOYSA-N
SMILES: COC1=CC=C(N=O)C(O)=C1
Percent Purity 
  • (2)
Physical Form 
  • (2)
Quantity 
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results