accessibility menu, dialog, popup

UserName

Viewing profile as user:
CH3CH3HNNClNNHH3CCH3N

CAS RN 139-40-2

IUPAC Name: 6-chloro-N2,N4-bis(propan-2-yl)-1,3,5-triazine-2,4-diamine
Synonyms: propazine prozinex
Molecular Weight (g/mol): 229.71
Molecular Formula: C9H16ClN5
InChi Key: WJNRPILHGGKWCK-UHFFFAOYSA-N
SMILES: CC(C)NC1=NC(NC(C)C)=NC(Cl)=N1

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results