accessibility menu, dialog, popup

UserName

Viewing profile as user:

CAS RN 139-45-7

IUPAC Name: 1,3-bis(propanoyloxy)propan-2-yl propanoate
Synonyms: tripropionin
Molecular Weight (g/mol): 260.29
Molecular Formula: C12H20O6
InChi Key: YZWRNSARCRTXDS-UHFFFAOYSA-N
SMILES: CCC(=O)OCC(COC(=O)CC)OC(=O)CC
Boiling Point 
  • (2)
  • (2)
Molecular Weight (g/mol) 
  • (2)
  • (2)
Percent Purity 
  • (2)
  • (2)
Quantity 
  • (1)
  • (1)
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (2)
  • (2)
  • (2)
  • (2)
  • (2)
  • (2)
  • (2)
  • (2)
  • (2)
  • (4)
  • (1)
  • (1)
  • (1)
  • (1)