accessibility menu, dialog, popup

UserName

Viewing profile as user:
NH2O

CAS RN 139-59-3

IUPAC Name: 4-phenoxyaniline
Synonyms: P-phenoxyaniline
Molecular Weight (g/mol): 185.23
Molecular Formula: C12H11NO
InChi Key: WOYZXEVUWXQVNV-UHFFFAOYSA-N
SMILES: NC1=CC=C(OC2=CC=CC=C2)C=C1
Molecular Weight (g/mol) 
  • (5)
  • (2)
Percent Purity 
  • (1)
  • (1)
  • (5)
Quantity 
  • (2)
  • (2)
  • (1)
  • (2)
Boiling Point 
  • (3)
  • (2)
  • (1)
Physical Form 
  • (3)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (6)
  • (1)
  • (6)
  • (5)
  • (2)
  • (1)
  • (1)
  • (5)
  • (2)
  • (2)
  • (1)
  • (2)
  • (3)
  • (2)
  • (1)
  • (3)
  • (1)