CAS RN 139015-32-0
CAS RN 139015-32-0
IUPAC Name:
N-methylpyrrolidin-3-amine
Synonyms:
N-methylpyrrolidin-3-amine
Molecular Weight (g/mol):
100.17
Molecular Formula:
C5H12N2
InChi Key:
NGZYRKGJWYJGRS-UHFFFAOYNA-N
SMILES:
CNC1CCNC1
1
–
1
of
1
results
1
(3S)-(-)-3-(Methylamino)pyrrolidine 97.0+%, TCI America™
CAS: 139015-32-0 Molecular Formula: C5H12N2 Molecular Weight (g/mol): 100.165 MDL Number: MFCD00191462 InChI Key: NGZYRKGJWYJGRS-YFKPBYRVSA-N PubChem CID: 10261057 IUPAC Name: (3S)-N-methylpyrrolidin-3-amine SMILES: CNC1CCNC1