accessibility menu, dialog, popup

UserName

Viewing profile as user:

CAS RN 139047-55-5

IUPAC Name: 2-[(4-phenylpiperazin-1-yl)methyl]-2H,3H,5H,6H-imidazo[1,2-c]quinazolin-5-one
Synonyms: 2-[(4-phenylpiperazin-1-yl)methyl]-2H,3H,6H-imidazo[1,2-c]quinazolin-5-one
Molecular Weight (g/mol): 361.45
Molecular Formula: C21H23N5O
InChi Key: ONSOQRPYFKDOEB-UHFFFAOYNA-N
SMILES: O=C1NC2=CC=CC=C2C2=NC(CN3CCN(CC3)C3=CC=CC=C3)CN12
Quantity 
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results