accessibility menu, dialog, popup

UserName

Viewing profile as user:
OHNSHN

CAS RN 13906-09-7

IUPAC Name: 2-sulfanylidene-1,2,3,4-tetrahydroquinazolin-4-one
Synonyms: 2-sulfanylidene-1,3-dihydroquinazolin-4-one
Molecular Weight (g/mol): 178.21
Molecular Formula: C8H6N2OS
InChi Key: PUPFOFVEHDNUJU-UHFFFAOYSA-N
SMILES: O=C1NC(=S)NC2=CC=CC=C12
Percent Purity 
  • (2)
Physical Form 
  • (2)
Quantity 
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (2)
  • (2)
  • (2)
  • (2)
  • (1)
  • (1)