accessibility menu, dialog, popup

UserName

Viewing profile as user:

CAS RN 13912-65-7

IUPAC Name: 3-{1,3-dioxo-1H,3H,3aH,4H,5H,9bH-naphtho[1,2-c]furan-5-yl}oxolane-2,5-dione
Synonyms: 3-{1,3-dioxo-3aH,4H,5H,9bH-naphtho[1,2-c]furan-5-yl}oxolane-2,5-dione
Molecular Weight (g/mol): 300.27
Molecular Formula: C16H12O6
InChi Key: JYCTWJFSRDBYJX-UHFFFAOYNA-N
SMILES: O=C1CC(C2CC3C(C(=O)OC3=O)C3=CC=CC=C23)C(=O)O1
Percent Purity 
  • (2)
Physical Form 
  • (2)
Quantity 
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (2)
  • (2)
  • (2)
  • (1)
  • (1)