accessibility menu, dialog, popup

UserName

Viewing profile as user:
CH3H3CNNCH3H3CCl

CAS RN 139143-09-2

IUPAC Name: 1,3-bis(propan-2-yl)-1H-imidazol-3-ium chloride
Synonyms: 1,3-diisopropylimidazol-1-ium chloride
Molecular Weight (g/mol): 188.7
Molecular Formula: C9H17ClN2
InChi Key: DOFXKPAOJLLPII-UHFFFAOYSA-M
SMILES: [Cl-].CC(C)N1C=C[N+](=C1)C(C)C
Molecular Weight (g/mol) 
  • (2)
  • (2)
Quantity 
  • (2)
  • (2)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (2)
  • (2)
  • (2)
  • (2)
  • (2)
  • (2)
  • (2)
  • (2)