accessibility menu, dialog, popup

UserName

Viewing profile as user:

CAS RN 139215-80-8

H3CFFF

CAS RN 139215-80-8

IUPAC Name: 4-ethyl-4'-(3,4,5-trifluorophenyl)-1,1'-bi(cyclohexane)
Synonyms: 4-ethyl-4'-(3,4,5-trifluorophenyl)-1,1'-bi(cyclohexane)
Molecular Weight (g/mol): 324.43
Molecular Formula: C20H27F3
InChi Key: DXFHMMVZUZLQFU-UHFFFAOYSA-N
SMILES: CCC1CCC(CC1)C1CCC(CC1)C1=CC(F)=C(F)C(F)=C1
Percent Purity 
  • (2)
Physical Form 
  • (2)
Quantity 
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results