accessibility menu, dialog, popup

UserName

Viewing profile as user:

CAS RN 13925-06-9

CH3H3CNNH3C

CAS RN 13925-06-9

IUPAC Name: 2-methyl-3-(2-methylpropyl)pyrazine
Synonyms: 2-methyl-3-(2-methylpropyl)pyrazine
Molecular Weight (g/mol): 150.23
Molecular Formula: C9H14N2
InChi Key: ZHMIODDNZRIENW-UHFFFAOYSA-N
SMILES: CC(C)CC1=NC=CN=C1C

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results