accessibility menu, dialog, popup

UserName

Viewing profile as user:
H3COOOOOOCH3CH3NNNH3C

CAS RN 1392500-07-0

IUPAC Name: 5-{2-azido-3-[(2,2-dimethyl-1,3-dioxan-5-yl)oxy]propoxy}-2,2-dimethyl-1,3-dioxane
Synonyms: 5-{2-azido-3-[(2,2-dimethyl-1,3-dioxan-5-yl)oxy]propoxy}-2,2-dimethyl-1,3-dioxane
Molecular Weight (g/mol): 345.4
Molecular Formula: C15H27N3O6
InChi Key: ZFDVYWOIFHYEAO-UHFFFAOYSA-N
SMILES: CC1(C)OCC(CO1)OCC(COC1COC(C)(C)OC1)N=[N+]=[N-]
Physical Form 
  • (1)
Quantity 
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results