accessibility menu, dialog, popup

UserName

Viewing profile as user:
OHNOOOHONHOOHO(E/Z)(E/Z)

CAS RN 139262-76-3

IUPAC Name: 3-[(4-{3-[4-(3-carboxyprop-2-enamido)phenoxy]phenoxy}phenyl)carbamoyl]prop-2-enoic acid
Synonyms: 3-[(4-{3-[4-(3-carboxyprop-2-enamido)phenoxy]phenoxy}phenyl)carbamoyl]prop-2-enoic acid
Molecular Weight (g/mol): 488.45
Molecular Formula: C26H20N2O8
InChi Key: HZFPOTBCYPWQSH-UHFFFAOYSA-N
SMILES: OC(=O)C=CC(=O)NC1=CC=C(OC2=CC(OC3=CC=C(NC(=O)C=CC(O)=O)C=C3)=CC=C2)C=C1

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results