accessibility menu, dialog, popup

UserName

Viewing profile as user:
NiH3CNCH3SSH3CNCH3SS

CAS RN 13927-77-0

IUPAC Name: nickel(2+) bis((dibutylcarbamothioyl)sulfanide)
Synonyms: nickel(2+) bis((dibutylcarbamothioyl)sulfanide)
Molecular Weight (g/mol): 467.43
Molecular Formula: C18H36N2NiS4
InChi Key: HPOWMHUJHHIQGP-UHFFFAOYSA-L
SMILES: [Ni++].CCCCN(CCCC)C([S-])=S.CCCCN(CCCC)C([S-])=S
Percent Purity 
  • (2)
Physical Form 
  • (2)
Quantity 
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results