accessibility menu, dialog, popup

UserName

Viewing profile as user:

CAS RN 139301-27-2

IUPAC Name: [4-(trifluoromethoxy)phenyl]boronic acid
Synonyms: 4-(trifluoromethoxy)phenylboronic acid
Molecular Weight (g/mol): 205.93
Molecular Formula: C7H6BF3O3
InChi Key: HUOFUOCSQCYFPW-UHFFFAOYSA-N
SMILES: OB(O)C1=CC=C(OC(F)(F)F)C=C1
Physical Form 
  • (4)
Quantity 
  • (1)
  • (1)
  • (2)
Percent Purity 
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (2)
  • (2)
  • (2)
  • (4)
  • (1)
  • (1)
  • (2)
  • (1)
  • (1)