accessibility menu, dialog, popup

UserName

Viewing profile as user:
PtNNNNClClH2O

CAS RN 13933-33-0

IUPAC Name: platinum(2+) tetraamine hydrate dichloride
Synonyms: platinum(2+) tetraamine hydrate dichloride
Molecular Weight (g/mol): 352.12
Molecular Formula: Cl2H14N4OPt
InChi Key: GWBDNMCYHWRSOH-UHFFFAOYSA-L
SMILES: N.N.N.N.O.[Cl-].[Cl-].[Pt++]
Physical Form 
  • (4)
Quantity 
  • (2)
  • (2)
Grade 
  • (2)
Percent Purity 
  • (2)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results