accessibility menu, dialog, popup

UserName

Viewing profile as user:

CAS RN 139332-64-2

H3CNCH3SOONNHNON

CAS RN 139332-64-2

IUPAC Name: N,N-dimethyl-7-(piperazin-1-yl)-2,1,3-benzoxadiazole-4-sulfonamide
Synonyms: N,N-dimethyl-7-(piperazin-1-yl)-2,1,3-benzoxadiazole-4-sulfonamide
Molecular Weight (g/mol): 311.36
Molecular Formula: C12H17N5O3S
InChi Key: XFQLOSBBYVMBGT-UHFFFAOYSA-N
SMILES: CN(C)S(=O)(=O)C1=CC=C(N2CCNCC2)C2=NON=C12
Percent Purity 
  • (1)
Physical Form 
  • (1)
Quantity 
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results