accessibility menu, dialog, popup

UserName

Viewing profile as user:

CAS RN 139494-68-1

IUPAC Name: sodium tris[(1,1,1,3,3,3-hexafluoropropan-2-yl)oxy]-λ²-boranuide
Synonyms: sodium tris[(1,1,1,3,3,3-hexafluoropropan-2-yl)oxy]-λ²-boranuide
Molecular Weight (g/mol): 534.89
Molecular Formula: C9H3BF18NaO3
InChi Key: FRRJTMMEYJQWIY-UHFFFAOYSA-N
SMILES: [Na+].FC(F)(F)C(O[B-](OC(C(F)(F)F)C(F)(F)F)OC(C(F)(F)F)C(F)(F)F)C(F)(F)F
Percent Purity 
  • (1)
Physical Form 
  • (1)
Quantity 
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results