accessibility menu, dialog, popup

UserName

Viewing profile as user:
BrNN

CAS RN 139585-70-9

IUPAC Name: 6-bromopyridine-3-carbonitrile
Synonyms: 6-bromopyridine-3-carbonitrile
Molecular Weight (g/mol): 183.01
Molecular Formula: C6H3BrN2
InChi Key: XHYGUDGTUJPSNX-UHFFFAOYSA-N
SMILES: BrC1=CC=C(C=N1)C#N
Percent Purity 
  • (2)
  • (2)
Physical Form 
  • (2)
  • (2)
Quantity 
  • (2)
  • (2)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (2)
  • (2)
  • (2)
  • (2)
  • (2)
  • (2)
  • (2)
  • (2)
  • (2)