accessibility menu, dialog, popup

UserName

Viewing profile as user:
NNOOOO

CAS RN 1395881-58-9

IUPAC Name: 17-{8,14-dioxa-1-azapentacyclo[11.7.1.0²,⁷.0⁹,²¹.0¹⁵,²⁰]henicosa-2(7),3,5,9,11,13(21),15,17,19-nonaen-5-yl}-8,14-dioxa-1-azapentacyclo[11.7.1.0²,⁷.0⁹,²¹.0¹⁵,²⁰]henicosa-2,4,6,9(21),10,12,15(20),16,18-nonaene
Synonyms: 17-{8,14-dioxa-1-azapentacyclo[11.7.1.0²,⁷.0⁹,²¹.0¹⁵,²⁰]henicosa-2(7),3,5,9,11,13(21),15,17,19-nonaen-5-yl}-8,14-dioxa-1-azapentacyclo[11.7.1.0²,⁷.0⁹,²¹.0¹⁵,²⁰]henicosa-2,4,6,9(21),10,12,15(20),16,18-nonaene
Molecular Weight (g/mol): 544.57
Molecular Formula: C36H20N2O4
InChi Key: QKKFYYGKAXHDOK-UHFFFAOYSA-N
SMILES: O1C2=CC=CC=C2N2C3=C(OC4=CC=CC1=C24)C=C(C=C3)C1=CC2=C(C=C1)N1C3=CC=CC=C3OC3=C1C(O2)=CC=C3
Physical Form 
  • (2)
Quantity 
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results