accessibility menu, dialog, popup

UserName

Viewing profile as user:

CAS RN 13961-36-9

IUPAC Name: 1-(prop-2-en-1-yl)piperazine
Synonyms: 1-(prop-2-en-1-yl)piperazine
Molecular Weight (g/mol): 126.2
Molecular Formula: C7H14N2
InChi Key: ZWAQJGHGPPDZSF-UHFFFAOYSA-N
SMILES: C=CCN1CCNCC1
Boiling Point 
  • (2)
  • (1)
Molecular Weight (g/mol) 
  • (1)
  • (2)
Percent Purity 
  • (2)
  • (1)
Quantity 
  • (1)
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (3)
  • (3)
  • (2)
  • (1)
  • (1)
  • (2)
  • (2)
  • (1)
  • (1)
  • (1)
  • (1)
  • (1)