accessibility menu, dialog, popup

UserName

Viewing profile as user:
BaOBrOOOBrOO

CAS RN 13967-90-3

IUPAC Name: barium(2+) dibromate
Synonyms: barium(2+) dibromate
Molecular Weight (g/mol): 393.13
Molecular Formula: BaBr2O6
InChi Key: VEASZGAADGZARC-UHFFFAOYSA-L
SMILES: [Ba++].[O-][Br](=O)=O.[O-][Br](=O)=O

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results