accessibility menu, dialog, popup

UserName

Viewing profile as user:

CAS RN 13972-68-4

IUPAC Name: cadmium(2+) molybdenum tetraoxidandiide
Synonyms: cadmium(2+) molybdenum tetraoxidandiide
Molecular Weight (g/mol): 272.36
Molecular Formula: CdMoO4
InChi Key: SQHWLBOPUKZGNS-UHFFFAOYSA-N
SMILES: [O--].[O--].[O--].[O--].[Mo].[Cd++]

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results