accessibility menu, dialog, popup

UserName

Viewing profile as user:
H3CCH3SiNNHNHNSiCH3H3CCH3CH3H3CH3CCH3CH3H3CH3CMg

CAS RN 1397288-30-0

IUPAC Name: magnesium(2+) 2,12-bis(2-phenylethynyl)-7,17-bis({2-[tris(propan-2-yl)silyl]ethynyl})-21,22,23,24-tetraazapentacyclo[16.2.1.1³,⁶.1⁸,¹¹.1¹³,¹⁶]tetracosa-4,14-diene-21,23-diide
Synonyms: magnesium(2+) 2,12-bis(2-phenylethynyl)-7,17-bis[2-(triisopropylsilyl)ethynyl]-21,22,23,24-tetraazapentacyclo[16.2.1.1³,⁶.1⁸,¹¹.1¹³,¹⁶]tetracosa-4,14-diene-21,23-diide
Molecular Weight (g/mol): 911.77
Molecular Formula: C58H78MgN4Si2
InChi Key: VHEGPZQOXOSWMG-UHFFFAOYNA-N
SMILES: [Mg++].CC(C)[Si](C#CC1C2CCC([N-]2)C(C#CC2=CC=CC=C2)C2NC(C=C2)C(C#C[Si](C(C)C)(C(C)C)C(C)C)C2CCC([N-]2)C(C#CC2=CC=CC=C2)C2NC1C=C2)(C(C)C)C(C)C
Percent Purity 
  • (1)
Physical Form 
  • (1)
Quantity 
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results