accessibility menu, dialog, popup

UserName

Viewing profile as user:
H3CNNH3CONNHOH3CSOONNCH3

CAS RN 139755-83-2

IUPAC Name: 5-{2-ethoxy-5-[(4-methylpiperazin-1-yl)sulfonyl]phenyl}-1-methyl-3-propyl-1H,4H,7H-pyrazolo[4,3-d]pyrimidin-7-one
Synonyms: revatio sildenafil viagra
Molecular Weight (g/mol): 474.58
Molecular Formula: C22H30N6O4S
InChi Key: BNRNXUUZRGQAQC-UHFFFAOYSA-N
SMILES: CCCC1=NN(C)C2=C1NC(=NC2=O)C1=CC(=CC=C1OCC)S(=O)(=O)N1CCN(C)CC1

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results