accessibility menu, dialog, popup

UserName

Viewing profile as user:
CH3NNNOOOH2NCH3

CAS RN 139756-01-7

IUPAC Name: 1-methyl-4-nitro-3-propyl-1H-pyrazole-5-carboxamide
Synonyms: 2-methyl-4-nitro-5-propylpyrazole-3-carboxamide
Molecular Weight (g/mol): 212.21
Molecular Formula: C8H12N4O3
InChi Key: BMLPAJIEDKJHSB-UHFFFAOYSA-N
SMILES: CCCC1=NN(C)C(C(N)=O)=C1[N+]([O-])=O
Percent Purity 
  • (2)
Physical Form 
  • (2)
Quantity 
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results