CAS RN 139756-21-1
IUPAC Name:
5-(2-ethoxyphenyl)-1-methyl-3-propyl-1H,4H,7H-pyrazolo[4,3-d]pyrimidin-7-one
Synonyms:
5-(2-ethoxyphenyl)-1-methyl-3-propyl-4H-pyrazolo[4,3-d]pyrimidin-7-one
Molecular Weight (g/mol):
312.37
Molecular Formula:
C17H20N4O2
InChi Key:
MXQUEDUMKWBYHI-UHFFFAOYSA-N
SMILES:
CCCC1=NN(C)C2=C1NC(=NC2=O)C1=CC=CC=C1OCC
1
–
1
of
1
results
1
5-(2-Ethoxyphenyl)-1-methyl-3-propyl-1,6-dihydro-7H-pyrazolo[4,3-d]-7-pyrimidinone 98.0+%, TCI America™
CAS: 139756-21-1 Molecular Formula: C17H20N4O2 Molecular Weight (g/mol): 312.37 MDL Number: MFCD03093929 InChI Key: MXQUEDUMKWBYHI-UHFFFAOYSA-N PubChem CID: 899745 IUPAC Name: 5-(2-ethoxyphenyl)-1-methyl-3-propyl-1H,4H,7H-pyrazolo[4,3-d]pyrimidin-7-one SMILES: CCCC1=NN(C)C2=C1NC(=NC2=O)C1=CC=CC=C1OCC